missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Ramiprilat-d3 ~90%(Mixture of Diastereomers), TRC
Specifications
Specifications
| Kemisk navn eller materiale | Ramiprilat-d3 |
| CAS | 1356933-70-4 |
| Molekylær formel | C21 2H3 H25 N2 O5 |
| Mængde | 1 mg |
| Synonym | (2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]-1-oxopropyl-d3]octahydrocyclopenta[b]pyrrole-2-carboxylic Acid, HOE 498 Diacid-d3, Ramipril Diacid-d3 |
| InChI formel | InChI=1S/C21H28N2O5/c1-13(22-16(20(25)26)11-10-14-6-3-2-4-7-14)19(24)23-17-9-5-8-15(17)12-18(23)21(27)28/h2-4,6-7,13,15-18,22H,5,8-12H2,1H3,(H,25,26)(H,27,28)/t13-,15-,16-,17-,18-/m0/s1/i1D3 |
| SMIL | [2H]C([2H])([2H])[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)N2[C@H]3CCC[C@H]3C[C@H]2C(=O)O |
| IUPAC navn | (2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]-3,3,3-trideuteriopropanoyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]pyrrole-2-carboxylic acid |
| Molekylvægt (g/mol) | 391.48 |
| Formel vægt | 391.2187 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?