Få mere at vide
Ramipril Diketopiperazine, TRC
Artikelnummer. 30135188
Skift visning
Mængde:
1 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135188 25 mg
30134203 1 mg
30006180 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135188 Leverandør Toronto Research Chemicals Leverandørnr. TRCR11100825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 108731-95-9
Molekylær formel C23 H30 N2 O4
Molekylvægt (g/mol) 398.5
Synonym Ethyl (2S)-2-[(3S,5aS,8aS,9aS)-3-Methyl-1,4-dioxodecahydro-2H-cyclopenta[4,5]pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate, Ramipril Imp. D (EP), Ramipril Diketopiperazine
IUPAC navn ethyl (2S)-2-[(2S,4aS,5aS,8aS)-2-methyl-1,4-dioxo-2,4a,5,5a,6,7,8,8a-octahydrocyclopenta[3,4]pyrrolo[3,5-c]pyrazin-3-yl]-4-phenylbutanoate
SMIL CCOC(=O)[C@H](CCc1ccccc1)N2[C@@H](C)C(=O)N3[C@H]4CCC[C@H]4C[C@H]3C2=O
Tekniske data
Kemisk navn eller materiale Ramipril Diketopiperazine
Mængde 25 mg
InChI formel InChI=1S/C23H30N2O4/c1-3-29-23(28)19(13-12-16-8-5-4-6-9-16)24-15(2)21(26)25-18-11-7-10-17(18)14-20(25)22(24)27/h4-6,8-9,15,17-20H,3,7,10-14H2,1-2H3/t15-,17-,18-,19-,20-/m0/s1
Formel vægt 398.2206