Få mere at vide
Ramipril-d5 (Major), TRC
Artikelnummer. 30418283
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30418283 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418283 Leverandør Toronto Research Chemicals Leverandørnr. TRCR11100425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ramipril-d5
CAS 1132661-86-9
Molekylær formel C23H27D5N2O5
Mængde 25 mg
Synonym (2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-(phenyl-d5)propyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic Acid, Altace-d5, Cardace-d5, Delix-d5, HOE-498-d5, Pramace-d5, Ramace-d5, Unipril-d5
InChI formel InChI=1S/C23H32N2O5/c1-3-30-23(29)18(13-12-16-8-5-4-6-9-16)24-15(2)21(26)25-19-11-7-10-17(19)14-20(25)22(27)28/h4-6,8-9,15,17-20,24H,3,7,10-14H2,1-2H3,(H,27,28)/t15-,17-,18-,19-,20-/m0/s1/i4D,5D,6D,8D,9D
SMIL C[C@@H](C(N1[C@](CCC2)([H])[C@]2([H])C[C@H]1C(O)=O)=O)N[C@H](C(OCC)=O)CCC3=C([2H])C([2H])=C([2H])C([2H])=C3[2H]
IUPAC navn (2S,3aS,6aS)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-(2,3,4,5,6-pentadeuteriophenyl)butan-2-yl]amino]propanoyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]pyrrole-2-carboxylic acid
Molekylvægt (g/mol) 421.54
Formel vægt 421.2625