Få mere at vide
Ramifenazone-d7, TRC
Artikelnummer. 30425239
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30425239 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425239 Leverandør Toronto Research Chemicals Leverandørnr. TRCR11050250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ramifenazone-d7
CAS 1330180-51-2
Renhedsgrad noter HPLC
Molekylær formel C14 D7 H12 N3 O
Mængde 50 mg
Synonym 1,2-Dihydro-1,5-dimethyl-4-[(1-methylethyl-d7)amino]-2-phenyl-3H-pyrazol-3-one, 4-(Isopropylamino-d7)antipyrine, 1-Phenyl-2,3-dimethyl-4-(isopropyl-d7) aminopyrazolone, 4-(Isopropylamino-d7)phenazone, 4-(Isopropyl-d7)amino- 2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, 4-Mono(isopropyl-d7)amino-1-phenyl- 2,3-dimethyl-5-pyrazolone, Isopirina-d7, (Isopropyl-d7)aminophenazone, Isopyrin-d7, Isopyrine-d7
InChI formel InChI=1S/C14H19N3O/c1-10(2)15-13-11(3)16(4)17(14(13)18)12-8-6-5-7-9-12/h5-10,15H,1-4H3/i1D3,2D3,10D
SMIL [2H]C([2H])([2H])C([2H])(NC1=C(C)N(C)N(C1=O)c2ccccc2)C([2H])([2H])[2H]
IUPAC navn 4-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-1,5-dimethyl-2-phenylpyrazol-3-one
Molekylvægt (g/mol) 252.363
Formel vægt 252.197
Procent renhed >95