Få mere at vide
Ramelteon Metabolite M-II-d3 (mixture of R and S at the hydroxy position), TRC
Artikelnummer. 30416275
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416275 1 mg
30421488 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416275 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1100621MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246812-22-5
Molekylær formel C16 D3 H18 N O3
Molekylvægt (g/mol) 278.36
Synonym Ramelteon Metabolite M-II-d3 (RS-hydroxy)
IUPAC navn 3,3,3-trideuterio-2-hydroxy-N-[2-[(8S)-2,6,7,8-tetrahydro-1H-cyclopenta[e]benzofuran-8-yl]ethyl]propanamide
SMIL [2H]C([2H])([2H])C(O)C(=O)NCC[C@@H]1CCc2ccc3OCCc3c12
Tekniske data
Kemisk navn eller materiale 2-Hydroxy-N-[2-[(8S)-1,6,7,8-tetrahydro-2H-cyclopenta[e]benzofuran-8-yl]ethyl]propanamide-d3
Mængde 1 mg
InChI formel InChI=1S/C16H21NO3/c1-10(18)16(19)17-8-6-12-3-2-11-4-5-14-13(15(11)12)7-9-20-14/h4-5,10,12,18H,2-3,6-9H2,1H3,(H,17,19)/t10?,12-/m0/s1/i1D3
Formel vægt 278.171