Få mere at vide
Raloxifene-d4 Hydrochloride, TRC
Artikelnummer. 30422377
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30422377 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422377 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1000021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Raloxifene-d4 Hydrochloride
CAS 1188263-47-9
Molekylær formel C28H24D4ClNO4S
Mængde 1 mg
Synonym [6-Hydroxy-2-(4-hydroxyphenyl)-benzo[b]thien-3-yl][4-[2-(1-piperidinyl)ethoxy]phenyl]methanone-d4 Hydrochloride, Keoxifene-d4, LY-139481-d4, Evista-d4
InChI formel InChI=1S/C28H27NO4S.ClH/c30-21-8-4-20(5-9-21)28-26(24-13-10-22(31)18-25(24)34-28)27(32)19-6-11-23(12-7-19)33-17-16-29-14-2-1-3-15-29;/h4-13,18,30-31H,1-3,14-17H2;1H/i16D2,17D2;
SMIL OC(C=C1)=CC=C1C(S2)=C(C(C3=CC=C(OC([2H])([2H])C([2H])([2H])N4CCCCC4)C=C3)=O)C5=C2C=C(O)C=C5.Cl
IUPAC navn (6-hydroxy-2-(4-hydroxyphenyl)benzo[b]thiophen-3-yl)(4-(2-(piperidin-1-yl)ethoxy-1,1,2,2-d4)phenyl)methanone hydrochloride
Molekylvægt (g/mol) 514.07
Formel vægt 513.17