Få mere at vide
Rafoxanide, MedChemExpress
Beskrivelse
Rafoxanide is an orally active salicylanilide anthelmintic agent. Rafoxanide is an antiparasitic agent and can be used for the control of infestation with Hemonchus species and Fasciola species in sheep and cattle.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Rafoxanide |
| Fysisk form | Solid |
| CAS | 22662-39-1 |
| Farve | Off-White |
| Mængde | 1 g |
| Renhedsgrad noter | Research |
| Molekylær formel | C19H11Cl2I2NO3 |
| Til brug med (applikation) | COVID-19-immunoregulation |
| Opløselighedsinformation | DMSO : ≥ 31 mg/mL (49.52 mM) |
| SMIL | O=C(NC1=CC=C(OC2=CC=C(Cl)C=C2)C(Cl)=C1)C3=CC(I)=CC(I)=C3O |
| Vis mere |