missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RAF1 (cRAF) [Y340D, Y341D], GST Tag Recombinant Protein
Human RAF1 (cRAF) [Y340D, Y341D], GST Tag Recombinant Protein
4380.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_002871, Y340D, Y341D |
---|---|
Gene Alias | RAF1 (cRAF) Y340D Y341D |
Species | Human |
Protein Tag | GST Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11583537
|
Invitrogen™
PV3805 |
10μg |
4380.00 DKK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10211073
|
Invitrogen™
PV4046 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468553
|
Invitrogen™
PR7307A |
100 μg |
13290.00 DKK
100µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RAF1 (cRAF) is a MAP kinase kinase kinase (MAP3K) which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated RAF1 can phosphorylate and activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylates and activates the serine⁄threonine specific protein kinases ERK1 and ERK2. The RAF1 Y340D Y341D mutant is constitutively active.Specifications
NP_002871, Y340D, Y341D | |
Human | |
Baculovirus |
RAF1 (cRAF) Y340D Y341D | |
GST Tag |
For Research Use Only. Not for use in diagnostic procedures.