Få mere at vide
Ractopamine-d6 Hydrochloride, TRC
Artikelnummer. 30418297
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30418297 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418297 Leverandør Toronto Research Chemicals Leverandørnr. TRCR0714021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ractopamine-d6 hydrochloride
CAS 1276197-17-1
Renhedsgrad noter HPLC
Molekylær formel C18 2H6 H17 N O3 . Cl H
Mængde 1 mg
Synonym Ractopamine D6 Hydrochloride, 4-[2-[[1,1,1,2,3,3-Hexadeuterio-4-(4-hydroxyphenyl)butan-2-yl]amino]-1-hydroxyethyl]phenol hydrochloride
InChI formel InChI=1S/C18H23NO3.ClH/c1-13(2-3-14-4-8-16(20)9-5-14)19-12-18(22)15-6-10-17(21)11-7-15;/h4-11,13,18-22H,2-3,12H2,1H3;1H/i1D3,2D2,13D;
SMIL Cl.[2H]C([2H])([2H])C([2H])(NCC(O)c1ccc(O)cc1)C([2H])([2H])Cc2ccc(O)cc2
IUPAC navn 4-[2,2,3,4,4,4-hexadeuterio-3-[[2-hydroxy-2-(4-hydroxyphenyl)ethyl]amino]butyl]phenol;hydrochloride
Molekylvægt (g/mol) 343.88
Formel vægt 343.1821
Procent renhed >95