Få mere at vide
Ractopamine D6 hydrochloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30328707
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30328707 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328707 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA16805010AL100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ractopamine-d6 hydrochloride
CAS 1276197-17-1
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C18 2H6 H17 N O3 . Cl H
Mængde 1 mL
Synonym Ractopamine D6 Hydrochloride, 4-[2-[[1,1,1,2,3,3-Hexadeuterio-4-(4-hydroxyphenyl)butan-2-yl]amino]-1-hydroxyethyl]phenol hydrochloride
InChI formel InChI=1S/C18H23NO3.ClH/c1-13(2-3-14-4-8-16(20)9-5-14)19-12-18(22)15-6-10-17(21)11-7-15;/h4-11,13,18-22H,2-3,12H2,1H3;1H/i1D3,2D2,13D;
SMIL Cl.[2H]C([2H])([2H])C([2H])(NCC(O)c1ccc(O)cc1)C([2H])([2H])Cc2ccc(O)cc2
IUPAC navn 4-[2,2,3,4,4,4-hexadeuterio-3-[[2-hydroxy-2-(4-hydroxyphenyl)ethyl]amino]butyl]phenol;hydrochloride
Molekylvægt (g/mol) 343.88
Formel vægt 343.1821 g/mol