missing translation for 'onlineSavingsMsg'
Få mere at vide

Ractopamine-d3 HCl (2-hydroxyethyl-1,1,2-d3) (mixture of diastereomeres), TRC

Artikelnummer. 30418007
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418007 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418007 Leverandør Toronto Research Chemicals Leverandørnr. TRCR7140115MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ractopamine 1-hydroxyethyl-1,2,2-d3 hydrochloride
CAS 1219794-72-5
Molekylær formel C18 2H3 H20 N O3 . Cl H
Mængde 5 mg
Synonym Ractopamine D3 (1-hydroxyethyl-1,2,2-D3) hydrochloride, Ractopamine D3 hydrochloride, Ractopamine-d3 hydrochloride, 4-Hydroxy-α-[[[3-(4-hydroxyphenyl)-1-methylpropyl]amino]methyl-d2]benzenemethanol-α-d1 hydrochloride, 4-[2-[[4-(4-Hydroxyphenyl)butan-2-yl]amino]-1-hydroxyethyl-1,2,2-d3]phenol hydrochloride, Ractopamine-D3 (1-hydroxyethyl-1,2,2-D3) hydrochloride, Ractopamine-D3 hydrochloride, Ractopamine-d3 hydrochloride, 4-Hydroxy-α-[[[3-(4-hydroxyphenyl)-1-methylpropyl]amino]methyl-d2]benzenemethanol-α-d1 hydrochloride, 4-[2-[[4-(4-Hydroxyphenyl)butan-2-yl]amino]-1-hydroxyethyl-1,2,2-d3]phenol hydrochloride
InChI formel InChI=1S/C18H23NO3.ClH/c1-13(2-3-14-4-8-16(20)9-5-14)19-12-18(22)15-6-10-17(21)11-7-15;/h4-11,13,18-22H,2-3,12H2,1H3;1H/i12D2,18D;
SMIL Cl.[2H]C([2H])(NC(C)CCc1ccc(O)cc1)C([2H])(O)c2ccc(O)cc2
IUPAC navn 4-[3-[[1,1,2-trideuterio-2-hydroxy-2-(4-hydroxyphenyl)ethyl]amino]butyl]phenol;hydrochloride
Molekylvægt (g/mol) 340.86
Formel vægt 340.1633
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.