Få mere at vide
Ractopamine-d3 HCl (2-hydroxyethyl-1,1,2-d3) (mixture of diastereomeres), CDN
Artikelnummer. 30321300
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321300 0.01 g
30319459 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321300 Leverandør CDN Leverandørnr. CDND68390.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ractopamine 1-hydroxyethyl-1,2,2-d3 hydrochloride
CAS 1219794-72-5
Molekylær formel C18 2H3 H20 N O3 . Cl H
Mængde 0.01 g
Synonym Ractopamine-D3 (1-hydroxyethyl-1,2,2-D3) hydrochloride, Ractopamine-D3 hydrochloride, Ractopamine-d3 hydrochloride, 4-Hydroxy-α-[[[3-(4-hydroxyphenyl)-1-methylpropyl]amino]methyl-d2]benzenemethanol-α-d1 hydrochloride, 4-[2-[[4-(4-Hydroxyphenyl)butan-2-yl]amino]-1-hydroxyethyl-1,2,2-d3]phenol hydrochloride
InChI formel InChI=1 S/C18H23NO3.ClH/c1-13(2-3-14-4-8-16(20)9-5-14)19-12-18(22)15-6-10-17(21)11-7-15;/h4-11,13,18-22 H,2-3,12H2,1H3;1 H/i12D2,18 D;
SMIL Cl.[2 H]C([2 H])(NC(C)CCc1ccc(O)cc1)C([2 H])(O)c2ccc(O)cc2
IUPAC navn 4-[3-[[1,1,2-trideuterio-2-hydroxy-2-(4-hydroxyphenyl)ethyl]amino]butyl]phenol;hydrochloride
Molekylvægt (g/mol) 340.86
Formel vægt 340.1633 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity