Få mere at vide
Raclopride-d5 Hydrochloride, TRC
Artikelnummer. 30427113
Skift visning
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427113 1 mg
30410749 10 mg
30421771 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427113 Leverandør Toronto Research Chemicals Leverandørnr. TRCR0708021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217623-85-2
Molekylær formel C15H15D5Cl2N2O3 • HCl
Molekylvægt (g/mol) 352.273646
Synonym 3,5-Dichloro-N-[[(2S)-1-(ethyl-d5)-2-pyrrolidinyl]methyl]-2-hydroxy-6-methoxybenzamide Hydrochloride, (-)-Raclopride-d5 Hydrochloride, FLA 870-d5
IUPAC navn 3,5-dichloro-2-hydroxy-6-methoxy-N-[[(2S)-1-(1,1,2,2,2-pentadeuterioethyl)pyrrolidin-2-yl]methyl]benzamide;hydrochloride
SMIL [2H]C([2H])([2H])C([2H])([2H])N1CCC[C@H]1CNC(=O)C2=C(C(=CC(=C2OC)Cl)Cl)O.Cl
Tekniske data
Kemisk navn eller materiale Raclopride-d5 Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C15H20Cl2N2O3.ClH/c1-3-19-6-4-5-9(19)8-18-15(21)12-13(20)10(16)7-11(17)14(12)22-2;/h7,9,20H,3-6,8H2,1-2H3,(H,18,21);1H/t9-;/m0./s1/i1D3,3D2;
Formel vægt 387.0932