Learn More
Hamilton™ Filtered Pre-Sterilized Racked Pipette Tips
Delivering outstanding accuracy and precision, guaranteed.
Brand: Hamilton™ 68007-13
Additional Details : Weight : 0.20000kg
Description
- Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
- Pre-sterilized
- 96 tips to a rack
- Standard retention
Specifications
10 μL | |
Filtered | |
Sterile | |
Polypropylene |