missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ RAC1/RAC2/RAC3/CDC42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599700
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Antibody detects endogenous levels of total Rac1/2/3.
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
Tekniske data
| RAC1/RAC2/RAC3/CDC42 | |
| Polyclonal | |
| Unconjugated | |
| Rac2 | |
| AI323801; AI452260; AI747189; AL023026; AU018915; CDC42; CDC42Hs; Cell division control protein 42 homolog; cell division cycle 42; cell division cycle 42 (GTP binding protein); cell division cycle 42 homolog; cell migration-inducing gene 5 protein; D5Ertd559e; dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)); dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)); EN-7; EN-7 protein; G25K; G25K GTP-binding protein; growth-regulating protein; GTP binding protein, 25kDa; Gx; HSPC022; MIG5; p21-Rac1; p21-Rac2; p21-Rac3; protein EN-7; RAC1; Rac-1; Rac1B; RAC2; RAC3; Ras-like protein TC25; RAS-related C3 botulinum substrate 1; RAS-related C3 botulinum substrate 2; RAS-related C3 botulinum substrate 3; ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 2; ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2); ras-related C3 botulinum toxin substrate 3; ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3); Ras-related C3 botulinum toxin substrate 3 (rho family, small GTP-binding protein Rac2); rho family, small GTP binding protein Rac1; rho family, small GTP binding protein Rac3; small G protein; small GTP binding protein CDC42; TC25; TC-25; TKS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12540, 170758, 19353, 19354, 363875, 366957, 5879, 5880, 5881, 64465, 688319, 998 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P15153, P60763, P60764, P60766, P60953, P63000, P63001, Q05144, Q6RUV5, Q8CFN2 | |
| Cdc42, Rac1, Rac2, Rac3 | |
| A synthesized peptide derived from human RAC1(Accession P63000), corresponding to amino acid residues M45-A95. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion