Få mere at vide
rac Zearalenone-d6 (major), TRC
Artikelnummer. 30451432
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30451432 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451432 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ2705031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Zearalenone-d6
CAS 1185236-04-7
Renhedsgrad noter HPLC
Molekylær formel C18H16D6O5
Mængde 1 mg
Synonym 10,10,11-Trideuterio-15,17-dihydroxy-11-(trideuteriomethyl)-12-oxabicyclo[12.4.0]octadeca-1(18),2,14,16-tetraene-7,13-dione, rac-Zearalenone-d6, rac-Zearalenone-D6 (10-hydroxy-6-oxo-1-undecenyl-9,9,10,11,11,11-D6), (3RS,11EZ)-3,4,5,6,9,10-Hexahydro-14,16-dihydroxy-3-(methyl-d3)-1H-2-benzoxacyclotetradecin-1,7(8H)-dione-3,4,4-d3, 6-(10-Hydroxy-6-oxo-1-undecenyl-9,9,10,11,11,11-d6)-β-resorcylic acid lactone
InChI formel InChI=1S/C18H22O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,20-21H,2,4-6,8-9H2,1H3/i1D3,6D2,12D
SMIL [2H]C([2H])([2H])C1([2H])OC(=O)c2c(O)cc(O)cc2C=CCCCC(=O)CCC1([2H])[2H]
IUPAC navn 10,10,11-trideuterio-15,17-dihydroxy-11-(trideuteriomethyl)-12-oxabicyclo[12.4.0]octadeca-1(14),2,15,17-tetraene-7,13-dione
Molekylvægt (g/mol) 324.4
Formel vægt 324.1844
Procent renhed >95