Få mere at vide
rac Zearalanone-d6, TRC
Artikelnummer. 30453150
Skift visning
Mængde:
0.25 mg
Artikelnummer. Mængde
30453150 0.25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453150 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ2704420.25MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Zearalanone-d6
CAS 1795020-90-4
Renhedsgrad noter HPLC
Molekylær formel C18H18D6O5
Mængde 0.25 mg
Synonym 3,4,5,6,9,10,11,12-Octahydro-14,16-dihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1,7(8H)-dione-d6, (RS)-Zearalanone-d6, (±)-Zearalanone-d6, DL-Zearalanone-d6, (±)-3,4,5,6,9,10,11,12-Octahydro-14,16-dihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1,7(8H)-dione-d6
InChI formel InChI=1S/C18H24O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h10-12,20-21H,2-9H2,1H3/i1D3,6D2,12D
SMIL O=C(C1=C(O)C=C(O)C=C1CCCCC2)OC([2H])(C([2H])([2H])[2H])C([2H])([2H])CCC2=O
IUPAC navn 10,10,11-trideuterio-15,17-dihydroxy-11-(trideuteriomethyl)-12-oxabicyclo[12.4.0]octadeca-1(18),14,16-triene-7,13-dione
Molekylvægt (g/mol) 326.42
Formel vægt 326.2
Procent renhed >95