Få mere at vide
rac Xanthoanthrafil-d3, TRC
Artikelnummer. 30449120
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30449120 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449120 Leverandør Toronto Research Chemicals Leverandørnr. TRCX5000021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Xanthoanthrafil-d3
CAS 1216710-83-6
Molekylær formel C19H20D3N3O6
Mængde 1 mg
Synonym N-[(3,4-Dimethoxyphenyl)methyl]-2-[(2-hydroxy-1-methylethyl)amino]-5-nitro-benzamide-d3, Benzamidenafil-d3
InChI formel InChI=1S/C19H23N3O6/c1-12(11-23)21-16-6-5-14(22(25)26)9-15(16)19(24)20-10-13-4-7-17(27-2)18(8-13)28-3/h4-9,12,21,23H,10-11H2,1-3H3,(H,20,24)/i1D3
SMIL [2H]C([2H])([2H])C(CO)Nc1ccc(cc1C(=O)NCc2ccc(OC)c(OC)c2)[N+](=O)[O-]
IUPAC navn N-[(3,4-dimethoxyphenyl)methyl]-5-nitro-2-[(1,1,1-trideuterio-3-hydroxypropan-2-yl)amino]benzamide
Molekylvægt (g/mol) 392.42
Formel vægt 392.1775