Få mere at vide
rac Viloxazine-d5 Hydrochloride, TRC
Artikelnummer. 30455262
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30455262 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30455262 Leverandør Toronto Research Chemicals Leverandørnr. TRCV3124421MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Viloxazine-d5 Hydrochloride
CAS 1276483-10-3
Molekylær formel C13H15D5ClNO3
Mængde 1 mg
Synonym (+/-)-2-[(o-Ethoxy-d5)phenoxymethyl]morpholine, Catatrol-d5, ICI 58834-d5, Vicilan-d5, Viloxazin-d5 Hydrochloride, Vivalan-d5, Vivarint-d5, Vivilan-d5
InChI formel InChI=1S/C13H19NO3.ClH/c1-2-15-12-5-3-4-6-13(12)17-10-11-9-14-7-8-16-11;/h3-6,11,14H,2,7-10H2,1H3;1H/i1D3,2D2;
SMIL [2H]C([2H])(C([2H])([2H])[2H])OC1=C(OCC2CNCCO2)C=CC=C1.Cl
Molekylvægt (g/mol) 278.79
Formel vægt 278.1446