Få mere at vide
rac-trans Sertraline-d3 Hydrochloride, TRC
Artikelnummer. 30414662
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30414662 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414662 Leverandør Toronto Research Chemicals Leverandørnr. TRCS28000410MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac-trans Sertraline-d3 Hydrochloride
CAS 1330180-66-9
Molekylær formel C17 D3 H14 Cl2 N . Cl H
Mængde 10 mg
Synonym (±)-trans-Sertraline-D3 (N-methyl-D3) Hydrochloride, rac-trans Sertraline-d3 hydrochloride, (±)-trans-Sertraline-D3 hydrochloride, (1R,4S)-rel-4-(3,4-Dichlorophenyl)-N-(methyl-d3)-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride, (±)-trans-Sertraline-d3 HCl (N-methyl-d3)
InChI formel InChI=1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17+;/m0./s1/i1D3;
SMIL Cl.[2H]C([2H])([2H])N[C@@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13
IUPAC navn rac-(1R,4S)-4-(3,4-dichlorophenyl)-N-(trideuteriomethyl)-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
Molekylvægt (g/mol) 345.709
Formel vægt 344.069