Få mere at vide
rac-trans Jasmonic Acid Methyl Ester, TRC
Artikelnummer. 30133302
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133302 50 mg
30007257 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133302 Leverandør Toronto Research Chemicals Leverandørnr. TRCJ21053050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 20073-13-6
Molekylær formel C13H20O3
Molekylvægt (g/mol) 224.3
Synonym (1R,2R)-rel-3-Oxo-2-(2Z)-2-penten-1-ylcyclopentaneacetic Acid Methyl Ester, (Z)-trans-(+/-)-3-Oxo-2-(2Z)-2-penten-1-ylcyclopentaneacetic Acid Methyl Ester, (+/-)-Methyl Jasmonate, [1alpha,2beta(Z)]-3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid Methyl Ester, Jasmoneige, Methyl DL-Jasmonate, Methyl dl-Jasmonate;
IUPAC navn methyl 2-[rac-(1R,2R)-3-oxo-2-[rac-(Z)-pent-2-enyl]cyclopentyl]acetate
SMIL CC\C=C/CC1C(CC(=O)OC)CCC1=O
Tekniske data
Kemisk navn eller materiale rac-trans Jasmonic Acid Methyl Ester
Mængde 50 mg
InChI formel InChI=1S/C13H20O3/c1-3-4-5-6-11-10(7-8-12(11)14)9-13(15)16-2/h4-5,10-11H,3,6-9H2,1-2H3/b5-4-/t10-,11-/m1/s1
Formel vægt 224.1412