Få mere at vide
rac trans-4-Cotinine Carboxylic Acid Methyl Ester, TRC
Artikelnummer. 30356332
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356332 50 mg
30357831 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30356332 Leverandør Toronto Research Chemicals Leverandørnr. TRCC72515550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac trans-4-Cotinine Carboxylic Acid Methyl Ester
CAS 135028-97-6
Molekylær formel C12H14N2O3
Mængde 50 mg
Synonym (2R,3R)-rel-1-Methyl-5-oxo-2-(3-pyridinyl)-3-pyrrolidinecarboxylic Acid Methyl Ester, trans-1-Methyl-5-oxo-2-(3-pyridinyl)-3-pyrrolidinecarboxylic Acid Methyl Ester
InChI formel InChI=1S/C12H14N2O3/c1-14-10(15)6-9(12(16)17-2)11(14)8-4-3-5-13-7-8/h3-5,7,9,11H,6H2,1-2H3/t9-,11-/m0/s1
SMIL COC(=O)[C@H]1CC(=O)N(C)[C@H]1c2cccnc2
IUPAC navn methyl (2R,3S)-1-methyl-5-oxo-2-pyridin-3-ylpyrrolidine-3-carboxylate
Molekylvægt (g/mol) 234.25
Formel vægt 234.1004