Få mere at vide
rac Tolterodine-d14 Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | rac Tolterodine-d14 Hydrochloride |
| CAS | 1246816-99-8 |
| Molekylær formel | C22H18D14ClNO |
| Mængde | 1 mg |
| Synonym | 2-[3-[Bis[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]-1-phenylpropyl]-4-methylphenol hydrochloride, (±)-Tolterodine-d14 hydrochloride, N,N-Di(isopropyl-d7)-3-(2-hydroxy-5-methylphenyl)-3-phenylpropanamine hydrochloride, N,N-Di(isopropyl-d7)-3-(2-hydroxy-5-methylphenyl)-3-phenylpropylamine hydrochloride, N,N-Di(isopropyl-d7)-N-[3-(2-hydroxy-5-methylphenyl)-3-phenylpropyl]amine hydrochloride, Racemic tolterodine-d14 hydrochloride, rac-Tolterodine-d14 hydrochloride, rac-Tolterodine-D14 (N,N-di(isopropyl-D7)) hydrochloride |
| InChI formel | InChI=1S/C22H31NO.ClH/c1-16(2)23(17(3)4)14-13-20(19-9-7-6-8-10-19)21-15-18(5)11-12-22(21)24;/h6-12,15-17,20,24H,13-14H2,1-5H3;1H |
| SMIL | Cl.[2H]C([2H])([2H])C([2H])(N(CCC(c1ccccc1)c2cc(C)ccc2O)C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H] |
| Molekylvægt (g/mol) | 376.03 |
| Formel vægt | 375.3051 |