Få mere at vide
rac Timolol Maleate, TRC
Artikelnummer. 30450436
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30450436 5 mg
30456372 25 mg
30453400 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30450436 Leverandør Toronto Research Chemicals Leverandørnr. TRCT4437005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57073-55-9
Molekylær formel C13 H24 N4 O3 S . C4 H4 O4
Molekylvægt (g/mol) 432.49
Synonym 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-, (2Z)-2-butenedioate (1:1), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-, (2Z)-2-butenedioate (1:1) (salt) (9CI), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-, (±)-, (Z)-2-butenedioate (1:1) (salt), 1,2,5-Thiadiazole, 2-propanol deriv., (±)-3-Morpholino-4-(3-tert-butylamino-2-hydroxypropoxy)-1,2,5-thiadiazole hydrogen maleate, (±)-Timolol maleate, NSC 312896, dl-Timolol maleate, 1-[(1,1-Dimethylethyl)amino]-3-[[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy]-propan-2-ol maleate, Racemic timolol maleate, rac-Timolol maleate
IUPAC navn (Z)-but-2-enedioic acid;1-(tert-butylamino)-3-[(4-morpholin-4-yl-1,2,5-thiadiazol-3-yl)oxy]propan-2-ol
SMIL CC(C)(C)NCC(O)COc1nsnc1N2CCOCC2.OC(=O)\C=C/C(=O)O
Tekniske data
Kemisk navn eller materiale rac Timolol Maleate
Mængde 5 mg
InChI formel InChI=1S/C13H24N4O3S.C4H4O4/c1-13(2,3)14-8-10(18)9-20-12-11(15-21-16-12)17-4-6-19-7-5-17;5-3(6)1-2-4(7)8/h10,14,18H,4-9H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formel vægt 432.1679