Få mere at vide
rac-Talinolol, TRC
Artikelnummer. 17927534
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17927534 50 mg
17909334 5 mg
17927254 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17927534 Leverandør Toronto Research Chemicals Leverandørnr. TRCT00500050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57460-41-0
Molekylær formel C20H33N3O3
Molekylvægt (g/mol) 363.49
Synonym Urea, N-cyclohexyl-N'-[4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-, Urea, N-cyclohexyl-N'-[4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-, (±)-, N-Cyclohexyl-N'-[4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]urea, (±)-Talinolol, 1-(4-Cyclohexylureidophenoxy)-2-hydroxy-3-tert-butylaminopropane, 1-[3-(3-Cyclohexylureido)phenoxy]-3-(tert-butylamino)-2-propanol, 1-[4-(Cyclohexylureido)phenoxy]-3-(tert-butylamino)-2-propanol, Cordanum, Racemic talinolol, Talinolol
IUPAC navn 1-[4-[3-(tert-butylamino)-2-hydroxypropoxy]phenyl]-3-cyclohexylurea
SMIL CC(C)(C)NCC(O)COc1ccc(NC(=O)NC2CCCCC2)cc1
Tekniske data
Kemisk navn eller materiale rac-Talinolol
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C20H33N3O3/c1-20(2,3)21-13-17(24)14-26-18-11-9-16(10-12-18)23-19(25)22-15-7-5-4-6-8-15/h9-12,15,17,21,24H,4-8,13-14H2,1-3H3,(H2,22,23,25)
Formel vægt 363.2522
Procent renhed >95%