missing translation for 'onlineSavingsMsg'
Learn More

rac Talinolol-d5, TRC

Product Code. 30413050
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30413050 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30413050 Supplier Toronto Research Chemicals Supplier No. TRCT00500210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale rac Talinolol-d5
CAS 1215343-94-4
Molekylær formel C20 2H5 H28 N3 O3
Mængde 10 mg
InChI formel InChI=1S/C20H33N3O3/c1-20(2,3)21-13-17(24)14-26-18-11-9-16(10-12-18)23-19(25)22-15-7-5-4-6-8-15/h9-12,15,17,21,24H,4-8,13-14H2,1-3H3,(H2,22,23,25)/i13D2,14D2,17D
SMIL [2H]C([2H])(NC(C)(C)C)C([2H])(O)C([2H])([2H])Oc1ccc(NC(=O)NC2CCCCC2)cc1
IUPAC navn 1-[4-[3-(tert-butylamino)-1,1,2,3,3-pentadeuterio-2-hydroxy-propoxy]phenyl]-3-cyclohexyl-urea
Molekylvægt (g/mol) 368.52
Formel vægt 368.2836
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.