Få mere at vide
rac styrenoxid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | rac Styrene Oxide |
| CAS | 96-09-3 |
| Molekylær formel | C8H8O |
| Mængde | 5 g |
| Synonym | 2-Phenyloxirane, (Epoxyethyl)benzene, 1,2-Epoxy-1-phenylethane, Phenyloxirane, (Epoxyethyl)benzene, (±)-Phenylethylene Oxide, (±)-Phenyloxirane, (±)-Styrene Oxide, 1,2-Epoxyethylbenzene, 1-Phenyl-1,2-epoxyethane, 2-Phenyloxirane, Epoxystyrene, NSC 637, NSC 7582, Phenethylene Oxide, Phenylethylene Oxide, Phenyloxirane, STO, Styrene 1',2'-Oxide, Styrene 7,8-Oxide, Styrene epoxide, Styryl Oxide, alpha,beta-Epoxystyrene |
| InChI formel | InChI=1S/C8H8O/c1-2-4-7(5-3-1)8-6-9-8/h1-5,8H,6H2 |
| SMIL | C1OC1c2ccccc2 |
| IUPAC navn | 2-phenyloxiran |
| Molekylvægt (g/mol) | 120.15 |
| Formel vægt | 120.0575 |