Få mere at vide
(Rac)-Sograzepide, MedChemExpress
Artikelnummer. 16465437
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
Pakningsstørrelse:
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16465437 50 mg 50mg
16424086 25 mg 25mg
16422578 5 mg -
16410379 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16465437 Leverandør Medchem Express Leverandørnr. HYU00360
MedChemExpress (Rac)-Sograzepide is an antagonist of cholecystokinin B (CCK-B) receptor, and has the potential of reducing the secretion of gastric acid.
(Rac)-Sograzepide is an antagonist of cholecystokinin B (CCK-B) receptor, and has the potential of reducing the secretion of gastric acid.
Tekniske data
Kemisk navn eller materiale (Rac)-Sograzepide
CAS 168161-71-5
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C28H30N6O3
Mængde 50 mg
Synonym (Rac)-Netazepide (Rac)-YF 476 (Rac)-YM-220
Opløselighedsinformation DMSO : 125 mg/mL (250.71 mM; Need ultrasonic)
SMIL O=C(NC1=CC=CC(NC)=C1)NC2C(N(CC(C(C)(C)C)=O)C3=CC=CC=C3C(C4=NC=CC=C4)=N2)=O
Molekylvægt (g/mol) 498.58
Formel vægt 498.58
Procent renhed 99.04%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation