Få mere at vide
rac Secoisolariciresinol-d6, TRC
Artikelnummer. 30423976
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30423976 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423976 Leverandør Toronto Research Chemicals Leverandørnr. TRCS23950310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Secoisolariciresinol-d6
CAS 1346603-90-4
Molekylær formel C20H20D6O6
Mængde 10 mg
Synonym (2R,3R)-rel-2,3-Bis[[4-hydroxy-3-methoxy(phenyl-d3)methyl]-1,4-butanediol, (R*,R*)-(±)-2,3-Bis[[4-hydroxy-3-methoxy(phenyl-d3)]methyl]-1,4-butanediol, (R*,R*)-2,3-Bis[[4-hydroxy-3-methoxy(phenyl-d3)]methyl]-1,4-butanediol
InChI formel InChI=1S/C20H26O6/c1-25-19-9-13(3-5-17(19)23)7-15(11-21)16(12-22)8-14-4-6-18(24)20(10-14)26-2/h3-6,9-10,15-16,21-24H,7-8,11-12H2,1-2H3/t15-,16-/m1/s1/i3D,4D,5D,6D,9D,10D
SMIL OC(C(OC)=C1[2H])=C([2H])C([2H])=C1C[C@H](CO)[C@@H](CO)CC2=C([2H])C(OC)=C(O)C([2H])=C2[2H]
IUPAC navn (2S,3S)-2,3-bis[(2,3,6-trideuterio-4-hydroxy-5-methoxyphenyl)methyl]butane-1,4-diol
Molekylvægt (g/mol) 368.45
Formel vægt 368.2106