missing translation for 'onlineSavingsMsg'
Få mere at vide

rac-Rotigotine-D3 Methylether, TRC

Artikelnummer. 30421000
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421000 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421000 Leverandør Toronto Research Chemicals Leverandørnr. TRCR70071725MG
 Begrænset vare
Forventet forsendelse: 09-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale rac-Rotigotine-D3 Methyl Ether
CAS 1246820-80-3
Molekylær formel C20H24D3NOS
Mængde 25 mg
Synonym N-Propyl-N-(1,2,3,4-tetrahydro-5-methoxy-2-naphthalenyl)-2-thiopheneethanamine-D3, 2-(N-Propyl-N-2-thienylethylamino)-5-methoxytetralin-D3, N 0724-D3, 5-Methoxy-N-(propyl-3,3,3-D3)-N-(2-(thiophen-2-yl)ethyl)-1,2,3,4-tetrahydronaphthalen-2-amine
InChI formel InChI=1S/C20H27NOS/c1-3-12-21(13-11-18-7-5-14-23-18)17-9-10-19-16(15-17)6-4-8-20(19)22-2/h4-8,14,17H,3,9-13,15H2,1-2H3/i1D3
SMIL [2H]C([2H])([2H])CCN(CCc1cccs1)C2CCc3c(C2)cccc3OC
IUPAC navn 5-methoxy-N-(2-thiophen-2-ylethyl)-N-(3,3,3-trideuteriopropyl)-1,2,3,4-tetrahydronaphthalen-2-amin
Molekylvægt (g/mol) 332.52
Formel vægt 332.2002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.