Få mere at vide
rac Rotigotine-d3 Hydrochloride Salt, TRC
Artikelnummer. 30415557
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415557 1 mg
30421643 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415557 Leverandør Toronto Research Chemicals Leverandørnr. TRCR7007021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1215846-20-0
Molekylær formel C19H23D3ClNOS
Molekylvægt (g/mol) 354.95
Synonym 5,6,7,8-Tetrahydro-6-[propyl-d3[2-(2-thienyl)ethyl]amino]-1-naphthalenol Hydrochloride, rac-N 0437-d3, rac-N 0923-d3, rac-Neupro-d3 Hydrochloride, rac-Rotigotine-d3 Hydrochloride, rac-SPM 962-d3
SMIL OC1=C(CCC(N(CCC([2H])([2H])[2H])CCC2=CC=CS2)C3)C3=CC=C1.Cl
Tekniske data
Kemisk navn eller materiale rac Rotigotine-d3 Hydrochloride Salt
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C19H25NOS.ClH/c1-2-11-20(12-10-17-6-4-13-22-17)16-8-9-18-15(14-16)5-3-7-19(18)21;/h3-7,13,16,21H,2,8-12,14H2,1H3;1H/i1D3;
Formel vægt 354.1612
Procent renhed >95%