Få mere at vide
rac Rivastigmine-d6, TRC
Artikelnummer. 30419978
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30419978 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419978 Leverandør Toronto Research Chemicals Leverandørnr. TRCR54100210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Rivastigmine-d6
CAS 194930-04-6
Molekylær formel C14H16D6N2O2
Mængde 10 mg
Synonym N-Ethyl-N-methylcarbamic Acid 3-[1-(Dimethylamino-d6)ethyl]phenyl Ester, Ethylmethylcarbamic Acid 3-[1-(Dimethylamino-d6)ethyl]phenyl Ester
InChI formel InChI=1S/C14H22N2O2/c1-6-16(5)14(17)18-13-9-7-8-12(10-13)11(2)15(3)4/h7-11H,6H2,1-5H3/i3D3,4D3
SMIL CCN(C)C(OC1=CC(C(C)N(C([2H])([2H])[2H])C([2H])([2H])[2H])=CC=C1)=O
Molekylvægt (g/mol) 256.37
Formel vægt 256.2058