Få mere at vide
rac-Pregabalin, TRC
Artikelnummer. 30136230
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136230 25 mg
30133835 50 mg
17991245 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136230 Leverandør Toronto Research Chemicals Leverandørnr. TRCP70480025MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 128013-69-4
Molekylær formel C8 H17 N O2
Molekylvægt (g/mol) 159.23
Synonym rac-Pregabalin, Hexanoic acid, 3-(aminomethyl)-5-methyl-, (±)-Pregabalin, 3-(Aminomethyl)-5-methylhexanoic acid
IUPAC navn 3-(aminomethyl)-5-methylhexanoic acid
SMIL CC(C)CC(CN)CC(=O)O
Tekniske data
Kemisk navn eller materiale rac-Pregabalin
InChI formel InChI=1S/C8H17NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7H,3-5,9H2,1-2H3,(H,10,11)
Mængde 25 mg
Formel vægt 159.1259