Få mere at vide
rac-Pregabalin, Mikromol™
Artikelnummer. 30051073
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051073 25 mg
17970526 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051073 Leverandør Mikromol Leverandørnr. MM1376.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
rac-Pregabalin
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac-Pregabalin
CAS 128013-69-4
Analyt- eller komponentnavne rac-Pregabalin
Forsendelsestilstand Room Temperature
Molekylær formel C8 H17 N O2
Certificeringer/compliance ISO 17025
Synonym rac-Pregabalin, Hexanoic acid, 3-(aminomethyl)-5-methyl-, 3-(Aminomethyl)-5-methylhexanoic acid, (+/-)-Pregabalin
InChI formel InChI=1S/C8H17NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7H,3-5,9H2,1-2H3,(H,10,11)
SMIL CC(C)CC(CN)CC(=O)O
IUPAC navn 3-(aminomethyl)-5-methylhexanoic acid
Molekylvægt (g/mol) 159.23
Mængde 25 mg
Formel vægt 159.1259 g/mol