Få mere at vide
rac Phenylephrine Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | rac Phenylephrine Hydrochloride |
| CAS | 154-86-9 |
| Molekylær formel | C9H14ClNO2 |
| Mængde | 25 g |
| Synonym | Benzenemethanol','3-hydroxy-α-[(methylamino)methyl]-','hydrochloride (1:1), Benzenemethanol','3-hydroxy-α-[(methylamino)methyl]-','hydrochloride (9CI), Benzenemethanol','3-hydroxy-α-[(methylamino)methyl]-','hydrochloride','(±)-, Benzyl alcohol','m-hydroxy-α-[(methylamino)methyl]-','hydrochloride','(±)- (8CI), (±)-Phenylephrine hydrochloride, 1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride, DL-Phenylephrine hydrochloride, NSC 14029, RS-phenylephrine hydrochloride, rac-Phenylephrine hydrochloride |
| InChI formel | InChI=1S/C9H13NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,9-12H,6H2,1H3;1H |
| SMIL | Cl.CNCC(O)c1cccc(O)c1 |
| IUPAC navn | 3-[1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride |
| Molekylvægt (g/mol) | 203.67 |
| Formel vægt | 203.0713 |