Få mere at vide
rac Penbutolol, TRC
Artikelnummer. 30011974
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011974 250 mg
17972265 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011974 Leverandør Toronto Research Chemicals Leverandørnr. TRCP220500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 36507-48-9
Molekylær formel C18 H29 N O2
Molekylvægt (g/mol) 291.43
Synonym rac-Penbutolol, 2-Propanol, 1-(2-cyclopentylphenoxy)-3-[(1,1-dimethylethyl)amino]-, (±)-Isopenbutolol, (±)-Penbutolol
IUPAC navn 1-(tert-butylamino)-3-(2-cyclopentylphenoxy)propan-2-ol
SMIL CC(C)(C)NCC(O)COc1ccccc1C2CCCC2
Tekniske data
Kemisk navn eller materiale rac Penbutolol
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C18H29NO2/c1-18(2,3)19-12-15(20)13-21-17-11-7-6-10-16(17)14-8-4-5-9-14/h6-7,10-11,14-15,19-20H,4-5,8-9,12-13H2,1-3H3
Formel vægt 291.2198
Procent renhed >95%