Få mere at vide
rac Penbutolol-d9, TRC
Artikelnummer. 30422879
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30422879 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422879 Leverandør Toronto Research Chemicals Leverandørnr. TRCP22050225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Penbutolol-d9
CAS 1346747-40-7
Molekylær formel C18H20D9NO2
Mængde 25 mg
Synonym 1-(2-Cyclopentylphenoxy)-3-[(1,1-dimethylethyl-d9)amino]-2-propanol, 1-(tert-Butylamino-d9)-3-(o-cyclopentylphenoxy)-2-propanol
InChI formel InChI=1S/C18H29NO2/c1-18(2,3)19-12-15(20)13-21-17-11-7-6-10-16(17)14-8-4-5-9-14/h6-7,10-11,14-15,19-20H,4-5,8-9,12-13H2,1-3H3/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])C(C([2H])([2H])[2H])(C([2H])([2H])[2H])NCC(COC1=CC=CC=C1C2CCCC2)O
IUPAC navn 1-(2-cyclopentylphenoxy)-3-((2-(methyl-d3)propan-2-yl-1,1,1,3,3,3-d6)amino)propan-2-ol
Molekylvægt (g/mol) 300.48
Formel vægt 300.28