Få mere at vide
rac-Paroxetine-d4 Hydrochloride, TRC
Artikelnummer. 30412080
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30412080 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412080 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2057531MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac-Paroxetine-d4 Hydrochloride
CAS 1217683-35-6
Renhedsgrad noter HPLC
Molekylær formel C19H17D4ClFNO3
Mængde 1 mg
Synonym rac-3-[(1,3-Benzod-d4-ioxol-5-yloxy)methyl]-4-(4-fluorophenyl-d4)piperidine Hydrochloride, rac-Aropax-d4, rac-Deroxat-d4, rac-Paxil-d4, rac-Seroxat-d4
InChI formel InChI=1S/C19H20FNO3.ClH/c20-15-3-1-13(2-4-15)17-7-8-21-10-14(17)11-22-16-5-6-18-19(9-16)24-12-23-18;/h1-6,9,14,17,21H,7-8,10-12H2;1H/t14-,17-;/m0./s1/i1D,2D,3D,4D;
SMIL FC(C([2H])=C1[2H])=C([2H])C([2H])=C1[C@H]2[C@H](COC3=CC=C(OCO4)C4=C3)CNCC2.Cl
IUPAC navn rac-(3R,4S)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(2,3,5,6-tetradeuterio-4-fluorophenyl)piperidine;hydrochloride
Molekylvægt (g/mol) 369.85
Formel vægt 369.1445
Procent renhed >95