missing translation for 'onlineSavingsMsg'
Få mere at vide

rac O-Acetyl Pseudoephedrine Hydrochloride, TRC

Artikelnummer. 30314914
Change view
Click to view available options
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30314914 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314914 Supplier Toronto Research Chemicals Supplier No. TRCA187635250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (1S,2S)-O-Acetylpseudoephedrine Hydrochloride
CAS 1630-34-8
Molekylær formel C12 H17 N O2 . Cl H
Mængde 250 mg
Synonym Benzenemethanol, α-[1-(methylamino)ethyl]-, acetate (ester), hydrochloride, [S-(R*,R*)]- (9CI), Pseudoephedrine, acetate (ester), hydrochloride, (+)- (8CI), (1S,2S)-O-Acetylpseudoephedrine hydrochloride, (1S,2S)-2-(Methylamino)-1-phenylpropyl acetate hydrochloride, (αS)-α-[(1S)-1-(Methylamino)ethyl]benzenemethanol acetate (ester) hydrochloride
InChI formel InChI=1S/C12H17NO2.ClH/c1-9(13-3)12(15-10(2)14)11-7-5-4-6-8-11;/h4-9,12-13H,1-3H3;1H/t9-,12+;/m0./s1
SMIL Cl.CN[C@@H](C)[C@@H](OC(=O)C)c1ccccc1
IUPAC navn [(1S,2S)-2-(methylamino)-1-phenylpropyl] acetate;hydrochloride
Molekylvægt (g/mol) 243.73
Formel vægt 243.1026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.