Få mere at vide
rac-Nicotine-d4, TRC
Artikelnummer. 30482155
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30482155 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482155 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4124272.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nicotine-D4
CAS 350818-69-8
Renhedsgrad noter HPLC
Molekylær formel C10 2H4 H10 N2
Mængde 2.5 mg
Synonym 5-(1-Methyl-2-pyrrolidinyl)pyridine-2,3,4,6-d4, rac-Nicotine-D4, racemic Nicotine-D4
InChI formel InChI=1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/i2D,4D,6D,8D
SMIL [2H]c1nc([2H])c(C2CCCN2C)c([2H])c1[2H]
IUPAC navn 2,3,4,6-tetradeuterio-5-(1-methylpyrrolidin-2-yl)pyridine
Molekylvægt (g/mol) 166.26
Formel vægt 166.1408
Procent renhed >95