Få mere at vide
rac-Nicotine-13CD3, TRC
Artikelnummer. 30481208
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30481208 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481208 Leverandør Toronto Research Chemicals Leverandørnr. TRCN41242450MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac-Nicotine methyl-13C,d3
CAS 909014-86-4
Molekylær formel 13C C9 2H3 H11 N2
Mængde 50 mg
Synonym 3-[1-(Methyl-13C-d3)-2-pyrrolidinyl]pyridine, racemic Nicotine (Methyl-13C-D3), rac-Nicotine-13CD3, rac-Nicotine-13C-D3 (13C-methyl-D3)
InChI formel InChI=1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/i1+1D3
SMIL [2H][13C]([2H])([2H])N1CCCC1c2cccnc2
IUPAC navn 3-[1-(trideuterio(1^{13}C)methyl)pyrrolidin-2-yl]pyridine
Molekylvægt (g/mol) 166.24
Formel vægt 166.1379