Få mere at vide
rac Nebivolol-d4 (Major), TRC
Artikelnummer. 30465207
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30465207 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465207 Leverandør Toronto Research Chemicals Leverandørnr. TRCN3879031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Nebivolol-d4 (Major)
CAS 1219407-55-2
Renhedsgrad noter HPLC
Molekylær formel C22 2H4 H21 F2 N O4
Mængde 1 mg
Synonym 2,2-Dideuterio-2-[[1,1-dideuterio-2-(6-fluorochroman-2-yl)-2-hydroxyethyl]amino]-1-(6-fluorochroman-2-yl)ethanol, rac Nevibolol-d4
InChI formel InChI=1S/C22H25F2NO4/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2/i11D2,12D2
SMIL [2H]C([2H])(NC([2H])([2H])C(O)C1CCc2cc(F)ccc2O1)C(O)C3CCc4cc(F)ccc4O3
IUPAC navn 2,2-dideuterio-2-[[1,1-dideuterio-2-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)-2-hydroxyethyl]amino]-1-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)ethanol
Molekylvægt (g/mol) 409.46
Formel vægt 409.2003
Procent renhed >95