Få mere at vide
rac N-Propyl-2-cyanimidopyrrolidine-5-acetic Acid, TRC
Artikelnummer. 30421917
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421917 5 mg
30412443 100 mg
30420287 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30421917 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8351505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1373869-89-6
Molekylær formel C10H15N3O2
Molekylvægt (g/mol) 209.24
Synonym Pramipexole Related Compound, (2S,5E)-5-(Cyanoimino)-1-propyl-2-pyrrolidineacetic Acid
IUPAC navn 2-[(2S,5E)-5-cyanoimino-1-propylpyrrolidin-2-yl]acetic acid
SMIL CCCN1C(CCC1=NC#N)CC(=O)O
Tekniske data
Kemisk navn eller materiale N-Propyl-2S-cyanimidopyrrolidine-5-acetic Acid
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C10H15N3O2/c1-2-5-13-8(6-10(14)15)3-4-9(13)12-7-11/h8H,2-6H2,1H3,(H,14,15)/b12-9+/t8-/m0/s1
Formel vægt 209.1164
Procent renhed >95%