Få mere at vide
rac N,O-Didesmethyl Venlafaxine-d3, TRC
Artikelnummer. 30379954
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30379954 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379954 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4415771MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac N,O-Didesmethyl Venlafaxine-d3
CAS 1189468-67-4
Molekylær formel C15H20D3NO2
Mængde 1 mg
Synonym 4-[1-(1-Hydroxycyclohexyl)-2-(methyl-d3-amino)ethyl]phenol
InChI formel InChI=1S/C15H23NO2/c1-16-11-14(12-5-7-13(17)8-6-12)15(18)9-3-2-4-10-15/h5-8,14,16-18H,2-4,9-11H2,1H3/i1D3
SMIL OC1(CCCCC1)C(CNC([2H])([2H])[2H])C2=CC=C(O)C=C2
Molekylvægt (g/mol) 252.37