Få mere at vide
rac N’-Nitrosonornicotine 5’-Acetate (Mixture of Diastereomers), TRC
Artikelnummer. 30475246
Skift visning
Mængde:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475246 2 mg
30478026 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30475246 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5349952MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac N'-Nitrosonornicotine 5'-Acetate (Mixture of Diastereomers)
CAS 68743-65-7
Molekylær formel C11H13N3O3
Mængde 2 mg
Synonym 1-Nitroso-5-(3-pyridinyl)-2-pyrrolidinol 2-Acetate, 5'-Acetoxy-N'-nitrosonornicotine
InChI formel InChI=1S/C11H13N3O3/c1-8(15)17-11-5-4-10(14(11)13-16)9-3-2-6-12-7-9/h2-3,6-7,10-11H,4-5H2,1H3
SMIL O=NN1C(CCC1C2=CC=CN=C2)OC(C)=O
IUPAC navn 1-nitroso-5-(pyridin-3-yl)pyrrolidin-2-yl acetate
Molekylvægt (g/mol) 235.24
Formel vægt 235.1