Få mere at vide
rac N-Formiminoglutamic Acid, TRC
Artikelnummer. 30133057
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133057 1 g
30011869 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133057 Leverandør Toronto Research Chemicals Leverandørnr. TRCF7355001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 943418-43-7
Molekylær formel C6H8N2O5
Molekylvægt (g/mol) 188.14
Synonym (R,S)-2-(Formylazo)pentanedioic Acid, D,L-2-(Formylazo)glutaric Acid
SMIL OC(=O)CC\C(=N/NC=O)\C(=O)O
Tekniske data
Kemisk navn eller materiale rac N-Formiminoglutamic Acid
InChI formel InChI=1S/C6H8N2O5/c9-3-7-8-4(6(12)13)1-2-5(10)11/h3-4H,1-2H2,(H,10,11)(H,12,13)
Mængde 1 g
Formel vægt 188.0433