Få mere at vide
rac N-Desmethyl Mephenytoin, TRC
Artikelnummer. 30353074
Skift visning
Mængde:
5 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353074 5 mg
30350032 250 mg
30362637 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30353074 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2920605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 631-07-2
Molekylær formel C11 H12 N2 O2
Molekylvægt (g/mol) 204.23
Synonym 2,4-Imidazolidinedione, 5-ethyl-5-phenyl-, Hydantoin, 5-ethyl-5-phenyl- (6CI,7CI,8CI), 5-Ethyl-5-phenyl-2,4-imidazolidinedione, (RS)-5-Ethyl-5-phenyl-2,4-imidazolidinedione, 5-Ethyl-5-phenylhydantoin, 5-Phenyl-5-ethylhydantoin, Desmethylmephenytoin, Ethylphenylhydantoin, NSC 150466, NSC 33388, Nirvanol, Normephenytoin
IUPAC navn 5-ethyl-5-phenylimidazolidine-2,4-dione
SMIL CCC1(NC(=O)NC1=O)c2ccccc2
Tekniske data
Kemisk navn eller materiale rac N-Desmethyl Mephenytoin
Mængde 5 mg
InChI formel InChI=1S/C11H12N2O2/c1-2-11(8-6-4-3-5-7-8)9(14)12-10(15)13-11/h3-7H,2H2,1H3,(H2,12,13,14,15)
Formel vægt 204.0899