Få mere at vide
rac N-Desmethyl Mephenytoin-D5 (Major), TRC
Artikelnummer. 30365060
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30365060 2.5 mg 2.5mg
30359085 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365060 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2920622.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 119458-27-4
Molekylær formel C11 2H5 H7 N2 O2
Molekylvægt (g/mol) 209.26
Synonym 2,4-Imidazolidinedione, 5-ethyl-5-(phenyl-d5)- (9CI), rac N-Desmethyl Mephenytoin-D5, 5-Ethyl-5-(phenyl-d5)hydantoin, 5-Ethyl-5-(pentadeuterophenyl)hydantoin, Nirvanol-d5, Normephenytoin D5 (phenyl D5), 5-(Phenyl-d5)-5-ethylhydantoin
IUPAC navn 5-ethyl-5-(2,3,4,5,6-pentadeuteriophenyl)imidazolidine-2,4-dione
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C2(CC)NC(=O)NC2=O
Tekniske data
Kemisk navn eller materiale rac N-Desmethyl Mephenytoin-D5 (Major)
Mængde 2.5 mg
InChI formel InChI=1S/C11H12N2O2/c1-2-11(8-6-4-3-5-7-8)9(14)12-10(15)13-11/h3-7H,2H2,1H3,(H2,12,13,14,15)/i3D,4D,5D,6D,7D
Formel vægt 209.1213