Få mere at vide
rac-N-Desmethyl Bedaquiline, TRC
Artikelnummer. 30011779
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011779 1 mg
30010532 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011779 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2313101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 654654-76-9
Molekylær formel C31H29BrN2O2
Molekylvægt (g/mol) 541.48
Synonym rac-(1R,2S)-1-(6-Bromo-2-methoxyquinolin-3-yl)-4-(methylamino)-2-(naphthalen-1-yl)-1-phenylbutan-2-ol, (alphaR,betaS)-rel-6-Bromo-2-methoxy-alpha-[2-(methylamino)ethyl]-alpha-1-naphthalenyl-beta-phenyl-3-quinolineethanol
SMIL CNCC[C@](O)([C@H](c1ccccc1)c2cc3cc(Br)ccc3nc2OC)c4cccc5ccccc45
Tekniske data
Kemisk navn eller materiale rac-N-Desmethyl Bedaquiline
Mængde 1 mg
InChI formel InChI=1S/C31H29BrN2O2/c1-33-18-17-31(35,27-14-8-12-21-9-6-7-13-25(21)27)29(22-10-4-3-5-11-22)26-20-23-19-24(32)15-16-28(23)34-30(26)36-2/h3-16,19-20,29,33,35H,17-18H2,1-2H3/t29-,31-/m1/s1