Få mere at vide
rac N-Benzyl Nebivolol-d4, TRC
Artikelnummer. 30333371
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30333371 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30333371 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2857321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac N-Benzyl Nebivolol-d4
CAS 1246814-48-1
Molekylær formel C29H27D4F2NO4
Mængde 1 mg
Synonym alpha,alpha'-[[(Phenylmethyl)imino]bis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol-d4
InChI formel InChI=1S/C29H31F2NO4/c30-22-8-12-26-20(14-22)6-10-28(35-26)24(33)17-32(16-19-4-2-1-3-5-19)18-25(34)29-11-7-21-15-23(31)9-13-27(21)36-29/h1-5,8-9,12-15,24-25,28-29,33-34H,6-7,10-11,16-18H2/i17D2,18D2
SMIL [2H]C([2H])(C(O)C1CCc2cc(F)ccc2O1)N(Cc3ccccc3)C([2H])([2H])C(O)C4CCc5cc(F)ccc5O4
IUPAC navn 2-[benzyl-[1,1-dideuterio-2-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)-2-hydroxyethyl]amino]-2,2-dideuterio-1-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)ethanol
Molekylvægt (g/mol) 499.58
Formel vægt 499.2472